EMAIL THIS PAGE TO A FRIEND
Nucleosides, nucleotides & nucleic acids
DNA or RNA oligonucleotide 2'-hydrazides for chemoselective click-type ligation with carbonyl compounds.
PMID 21888548
Abstract
An efficient method for the synthesis of DNA or RNA oligonucleotide 2'-hydrazides is described. Fully deprotected oligonucleotides containing a hydrazide group at the 2'-position of a uridine residue were obtained by a novel two-step procedure: periodate cleavage of an oligonucleotide with 1,2-diol group followed by conversion of the aldehyde to hydrazide with an extended linker arm using a homobifunctional reagent succinic dihydrazide and NaBH(3)CN. The resulting oligonucleotide 2'-hydrazides were efficiently conjugated by a click-type reaction at acidic pH to aliphatic or aromatic aldehydes with or without NaBH(3)CN reduction to afford novel 2'-conjugates.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
S5502
Succinic dihydrazide, 96%
C4H10N4O2