EMAIL THIS PAGE TO A FRIEND
Analytical and bioanalytical chemistry
Ca(2+)-triggered coelenterazine-binding protein from Renilla as an enzyme-dependent label for binding assay.
PMID 21892640
Abstract
The recombinant Ca(2+)-triggered coelenterazine-binding protein (CBP) from Renilla muelleri was investigated as a biospecifically labeled molecule for in vitro assay applications. The protein was shown to be stable in solutions in the frozen state, as well as stable under heating and to chemical modifications. Conjugates with biotin, oligonucleotide, and proteins were obtained and applied as biospecific molecules in a solid-phase microassay. CBP detection was performed with intact (no modifications were made) Renilla luciferase in the presence of calcium, and the detection limit was found to be 75 amol. Model experiments indicate that this approach shows much promise, especially with regard to the development of multianalytical systems.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C2230
Coelenterazine, native, solid
C26H21N3O3