EMAIL THIS PAGE TO A FRIEND
Journal of industrial microbiology & biotechnology
Screening, cultivation, and biocatalytic performance of Rhodococcus boritolerans FW815 with strong 2,2-dimethylcyclopropanecarbonitrile hydratase activity.
PMID 21892773
Abstract
In this work, a mild, efficient bioconversion of 2,2-dimethylcyclopropanecarbonitrile (DMCPCN) to 2,2-dimethylcyclopropanecarboxamide (DMCPCA) in distilled water system was developed. The isolate FW815 was screened using the enrichment culture technique, displaying strong DMCPCN hydratase activity, and was identified as Rhodococcus boritolerans based on morphological, physiological, biochemical tests and 16S rRNA gene sequencing. Cultivation outcomes indicated that R. boritolerans FW815 was a neutrophile, with a growth optimum of 28-32°C; its DMCPCN hydratase belonged to the Fe-type family, and was most active at 38-42°C, pH 7.0, with maximal activity of 4.51 × 10(4) U g(-1) DCW. R. boritolerans FW815 was found to be DMCPCA amidase-negative, eliminating the contamination of dimethylcyclopropanecarboxylic acid. Moreover, it displayed high activity and acceptable reusability in the non-buffered distilled water system, comparable to those in pH 7.0 phosphate buffer (50.0 mmol l(-1)).