EMAIL THIS PAGE TO A FRIEND
Organic & biomolecular chemistry
Inhibition of peroxynitrite- and peroxidase-mediated protein tyrosine nitration by imidazole-based thiourea and selenourea derivatives.
PMID 21894311
Abstract
In the present study, the synthesis and characterization of a series of N-methylimidazole-based thiourea and selenourea derivatives are described. The new compounds were also studied for their ability to inhibit peroxynitrite (PN)- and peroxidase-mediated nitration of protein tyrosine residues. It has been observed that the selenourea derivatives are more efficient than the thiourea-based compounds in the inhibition of protein nitration. The higher activity of selenoureas as compared to that of the corresponding thioureas can be ascribed to the zwitterionic nature of the selenourea moiety. Single crystal X-ray diffraction studies on some of the thiourea and selenourea derivatives reveal that the C=S bonds in thioureas possess more of double bond character than the C=Se bonds in the corresponding selenoureas. Therefore, the selenium compounds can react with PN or hydrogen peroxide much faster than their sulfur analogues. The reactions of thiourea and selenourea derivatives with PN or hydrogen peroxide produce the corresponding sulfinic or seleninic acid derivatives, which upon elimination of sulfurous/selenous acids produce the corresponding N-methylimdazole derivatives.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M50834
1-Methylimidazole, ReagentPlus®, 99%
C4H6N2
230499
Selenourea, 98%
CH4N2Se