EMAIL THIS PAGE TO A FRIEND
American journal of clinical dermatology
These dermatological disorders share the features of being caused by hyperactivity of androgen within the pilosebaceous follicle, and although not life threatening are the source of a considerable psychological and emotional burden for patients. Foreword.
PMID 21895043
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C5145
Chlormadinone acetate, ≥98%
C23H29ClO4