EMAIL THIS PAGE TO A FRIEND
Nanoscale
Multifunctional nanoadditives for the thermodynamic and kinetic stabilization of enzymes.
PMID 21897968
Abstract
Stabilization of enzymes has become a major focus in the quest to improve the activity, sustainability and recyclability of enzymes for their successful integration into both industry and medicine. Here, we describe the kinetic and thermodynamic stabilization of a variety of enzymes in the presence of cationic multifunctional polymeric nanoparticles.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
705314
Poly(glycidyl methacrylate), Mn 10,000-20,000