EMAIL THIS PAGE TO A FRIEND
Chemistry (Weinheim an der Bergstrasse, Germany)
In search of oligo(2-thienyl)-substituted pyridine derivatives: a modular approach to di-, tri- and tetra(2-thienyl)pyridines.
PMID 21898622
Abstract
Herein, we describe our attempts to systematically prepare a series of oligo(2-thienyl)-substituted pyridine derivatives. The crucial starting material, a β-alkoxy-β-ketoenamide, is easily available on a large scale by the reaction of lithiated methoxyallene with thiophene-2-carbonitrile and thiophene-2-carboxylic acid. This three-component reaction is followed by intramolecular cyclization to yield the suitably functionalized 2,6-di(2-thienyl)-substituted pyridine derivates. The two oxygen atoms allow the programmed activation of positions C-3, C-4, or C-5 of the pyridine ring to perform palladium-catalyzed coupling reactions with thiophene-2-boronic acid or 2-(tributylstannyl)thiophene, and alternatively, reductive removal of groups. With this concept, we were able to prepare five pyridine derivatives with 2-thienyl substituents in the 2,6-, 2,3,6-, 2,4,6-, 2,3,4,6-, and 2,3,5,6-positions. 2,3,4,5,6-Penta(2-thienyl)pyridine was not available with our methods. The UV/Vis and fluorescence spectra of all pyridines were recorded and showed a dependence on the substitution pattern and protonation state. For the protonated 2,3,5,6-tetra(2-thienyl)-substituted pyridine, a Stokes shift of about 180 nm with an emission at 515 nm was observed.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T32603
2-Thiophenecarboxylic acid, ReagentPlus®, 99%
C5H4O2S