EMAIL THIS PAGE TO A FRIEND
Nihon Naika Gakkai zasshi. The Journal of the Japanese Society of Internal Medicine
[Series: Clinical study from Japan and its reflections; J-BAF study].
PMID 21899170
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B5016
Bepridil hydrochloride, powder
C24H34N2O · HCl