EMAIL THIS PAGE TO A FRIEND
The Journal of organic chemistry
Direct, metal-free amination of heterocyclic amides/ureas with NH-heterocycles and N-substituted anilines in POCl3.
PMID 21899258
Abstract
A POCl(3)-mediated, direct amination reaction of heterocyclic amides/ureas with NH-heterocycles or N-substituted anilines is described. Compared to the existing methods, this operationally simple protocol provides unique reactivity and functional group compatibility because of the metal-free, acidic reaction conditions. The yields are generally excellent.
Related Materials