EMAIL THIS PAGE TO A FRIEND
Organic letters
Expedient Route to the functionalized calyciphylline A-type skeleton via a Michael addition-RCM strategy.
PMID 21899283
Abstract
An efficient, robust, and scalable strategy to access the functionalized core of calyciphylline A-type alkaloids has been developed starting from commercially available 3-methylanisole. Key features of this approach are an intramolecular Michael addition/allylation sequence and a ring-closing metathesis step.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
149160
3-Methylanisole, 99%
C8H10O