EMAIL THIS PAGE TO A FRIEND
ACS nano
Evolution and protein packaging of small-molecule RNA aptamers.
PMID 21899290
Abstract
A high-affinity RNA aptamer (K(d) = 50 nM) was efficiently identified by SELEX against a heteroaryldihydropyrimidine structure, chosen as a representative drug-like molecule with no cross reactivity with mammalian or bacterial cells. This aptamer, its weaker-binding variants, and a known aptamer against theophylline were each embedded in a longer RNA sequence that was encapsidated inside a virus-like particle by a convenient expression technique. These nucleoprotein particles were shown by backscattering interferometry to bind to the small-molecule ligands with affinities similar to those of the free (nonencapsidated) aptamers. The system therefore comprises a general approach to the production and sequestration of functional RNA molecules, characterized by a convenient label-free analytical technique.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
745774
8-Amino-1-octanethiol hydrochloride, 95%
C8H19NS · HCl