EMAIL THIS PAGE TO A FRIEND
Journal of neurochemistry
Inhibition of neuronal cholesterol biosynthesis with lovastatin leads to impaired synaptic vesicle release even in the presence of lipoproteins or geranylgeraniol.
PMID 21899539
Abstract
Cholesterol is highly enriched in the brain, and plays a key role in synapse formation and function. The brain does not derive cholesterol from the circulation; instead, the majority of cholesterol is made in glia and secreted in form of lipoproteins. Neurons can synthesize cholesterol, but the extent of neuronal cholesterol biosynthesis in the adult brain is unknown. Cholesterol biosynthesis inhibitors of the statin family are widely used to lower circulating cholesterol and cardiovascular risk. Lipophilic statins can cross the blood brain barrier and inhibit brain cholesterol biosynthesis with possible consequences for synaptic cholesterol homeostasis. We have investigated the effects of lovastatin on synapse maturation and synaptic vesicle release. Treatment of primary hippocampal neurons with low levels of lovastatin for one week reduced synapse density and impaired synaptic vesicle release. Neither lipoproteins nor geranylgeraniol fully counteracted the lovastatin-induced decrease of synaptic vesicle exocytosis, even when cholesterol depletion was prevented. In contrast, restoration of neuronal cholesterol synthesis with mevalonate prevented defects in vesicle exocytosis without fully normalizing neuronal cholesterol content. These results raise the possibility that chronic exposure of neurons to lipophilic statins may affect synaptic transmission, and indicate that hippocampal neurons need a certain level of endogenous cholesterol biosynthesis.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
G3278
Geranylgeraniol, ≥85% (GC)
C20H34O