EMAIL THIS PAGE TO A FRIEND
Organic letters
Organocatalytic asymmetric aldol reaction of hydroxyacetone with β,γ-unsaturated α-keto esters: facile access to chiral tertiary alcohols.
PMID 21902199
Abstract
An efficient direct asymmetric aldol reaction between hydroxyacetone and β,γ-unsaturated α-keto esters has been successfully developed. In the presence of 9-amino-9-deoxy-epi-cinchonine and trifluoroacetic acid, the direct aldol reaction of O-protected hydroxyacetone proceeded in a highly enantioselective manner, affording the desired adducts containing a chiral tertiary alcohol in high yields and with excellent enantioselectivities. The aldol products obtained are valuable precursors for the synthesis of 2-substituted glycerol derivatives.
Related Materials