EMAIL THIS PAGE TO A FRIEND
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy
Synthesis and characterization of cadmium selenide nanoparticles loaded on activated carbon and its efficient application for removal of muroxide from aqueous solution.
PMID 21903452
Abstract
In the first, Cadmium selenide Nanoparticle loaded on activated carbon (CdSe-NP-AC) has been synthesized and characterized by different techniques including XRD and SEM. Then, this new adsorbent successfully has been applied for the removal of muroxide (MO) from aqueous solution in batch studies, while the effect of various experimental parameters like initial pH (pH(0)), contact time, amount of (CdSe-NP-AC) and initial MO concentration (C(0)) on its removal percentage was examined by one at a time optimization method. It was found following optimization of variable, the adsorption of MO onto (CdSe-NP-AC) followed pseudo-second-order kinetics and show Tempkin and Langmuir models for interpretation of experimental data. It was observed that by increasing the temperature the removal percentage was improved and the positive change in entropy (ΔS°) and heat of adsorption (ΔH°) show the endothermic nature of process, while the high negative value in Gibbs free energy change (ΔG°) indicates the feasible nature of adsorption process.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
222461
Murexide, ACS reagent
C8H8N6O6