EMAIL THIS PAGE TO A FRIEND
Angewandte Chemie (International ed. in English)
Identification of acyl protein thioesterases 1 and 2 as the cellular targets of the Ras-signaling modulators palmostatin B and M.
PMID 21905186
Abstract
Finding the target: activity-based proteomic profiling probes based on the depalmitoylation inhibitors palmostatin B and M have been synthesized and were found to target acyl protein thioesterase 1 (APT1) and 2 (APT2) in cells.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P5648
β-Propiolactone, Grade II, ≥90%
C3H4O2