EMAIL THIS PAGE TO A FRIEND
Journal of agricultural and food chemistry
Capillary electrophoresis of free fatty acids by indirect ultraviolet detection: application to the classification of vegetable oils according to their botanical origin.
PMID 21905655
Abstract
A method for the determination of fatty acids in vegetable oils by capillary electrophoresis with indirect UV-vis detection has been developed. The separation of fatty acids was optimized in terms of Brij surfactant nature and concentration and organic modifier (2-propanol) percentage. The optimal background electrolyte consisted of 10 mM p-hydroxybenzoate, 5 mM Tris at pH 8.8, 80 mM Brij 98, 40% acetonitrile, and 10% 2-propanol. Under these conditions, vegetable oils from five botanical origins (avocado, corn, extra virgin olive, hazelnut, and soybean) were analyzed and the fatty acid contents established. Linear discriminant analysis (LDA) models were constructed using fatty acid peak areas as predictors. An excellent resolution among all category pairs was obtained, and all samples were correctly classified with assignment probabilities of >95%.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
388866
Brij® 93, average Mn ~357
C22H44O3
P6136
Brij® O10
C38H76O11
436240
BRIJ® O20, average Mn ~1,150
C58H116O21