EMAIL THIS PAGE TO A FRIEND
The Journal of organic chemistry
Quinoxalino[2,3-c]cinnolines and their 5-N-oxide: alkoxylation of methyl-substituted quinoxalino[2,3-c]cinnolines to acetals and orthoesters.
PMID 21905671
Abstract
We report the alkoxylation of methyl-substituted quinoxalino[2,3-c]cinnolines to give acetals and orthoesters in high yields. Routes to the precursors of this alkoxylation reaction as well as other quinoxalino[2,3-c]cinnoline and their 5-oxide derivatives are reported. Most of these quinoxalino[2,3-c]cinnolines were prepared by cyclization of the corresponding 2-amino-3-(2-nitrophenyl)quinoxaline, which, in turn, result from an unusual Beirut reaction from benzofurazan oxides plus 2-nitrobenzylcyanides. Mechanistic explanations for these intriguing reactions are presented.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
650137
Benzofurazan, 97%
C6H4N2O
SML0552
Tirapazamine, ≥98% (HPLC)
C7H6N4O2