EMAIL THIS PAGE TO A FRIEND
Journal of dermatological science
Integral hair lipid in human hair follicle.
PMID 21906914
Abstract
Integral hair lipid (IHL) is bound to the keratinized cell surface to make an environmentally resistant lipid envelope. It is mainly positioned on the hair cuticle and inner root sheath. IHL in the hair follicle may regard as hair barrier to be similar to the epidermal lipid layer functioning as skin barrier. Major constituents of IHL are fatty acid, phytosphingosine, ceramide in decreasing order. Minor constituents of IHL are cholesterol, cholesterol sulfate and cholesterol oleate. Cuticle or cortical cell surface in hair are abundant in fatty acids unlike the keratinized area of epidermis or sebaceous gland, and about 30-40% of such fatty acids are composed of 18-methyl-eicosanoic acid which is known to be bound to proteins by ester or thioester bond. Various factors including moisture, solvent, oxidative damage during bleaching or permanent waving affect IHL. Photochemical changes also can occur in IHL as well as in hair protein and hair pigment. Lipid metabolism is thought to play an essential role in lipid envelope of hair, but also involvement in hair development and function.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C9523
Sodium cholesteryl sulfate
C27H45NaO4S