EMAIL THIS PAGE TO A FRIEND
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences
Analysis of cantharidin in false blister beetles (Coleoptera: Oedemeridae) by headspace solid-phase microextraction and gas chromatography-mass spectrometry.
PMID 21907641
Abstract
A headspace solid-phase microextraction (HS-SPME) coupled to gas chromatography-mass spectrometry (GC-MS) method was developed to determine a type of terpenoid named as cantharidin in the false blister beetles, family Oedemeridae. The experimental parameters for HS-SPME method were optimized. Six commercial fibers for HS-SPME method development were tested and the divinylbenzene/carboxene/polydimethylsiloxane fiber was selected to provide the best detection of analyzed compound. The calibration curve showed linearity in the range of 0.1-50 μg mL(-1), correlation coefficient (R(2)=0.992), limit of detection (0.01 ng mL(-1)) and quantitation (0.04 ng mL(-1)) were obtained for the proposed method. The relative standard deviations of intra-day and inter-day assays were 7.8 and 3.4%, respectively. The recovery values, obtained after spiking the beetle samples by three concentration levels of standard solution, were higher than 87%. The results indicated the successful application of the proposed method on the analysis of cantharidin from the false blister beetles.