EMAIL THIS PAGE TO A FRIEND
Carbohydrate research
Complete sets of monosubstituted cyclomaltohexaoses (α-cyclodextrins) as precursors for further synthesis.
PMID 21907975
Abstract
Alkylation of cyclomaltohexaose (α-cyclodextrin, α-CD) with allyl or cinnamyl bromide, followed by peracetylation of remaining hydroxyl groups and separation of isomers, resulted in the set of peracetylated 2(I)-O-, 3(I)-O- and 6(I)-O-alkylated α-CDs in up to 27% yields. Ozonolysis or oxidative cleavage of peracetylated allyl or cinnamyl derivatives resulted in a complete set of peracetylated 2(I)-O-, 3(I)-O- and 6(I)-O-formylmethyl or carboxymethyl derivatives that are useful precursors for preparation of regioselectively monosubstituted derivatives of α-CD. Moreover, a quick method to recognize single 2(I)-O-, 3(I)-O- and 6(I)-O-monosubstituted peracetylated CDs from one another using only their (1)H NMR spectra has been proposed.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
208523
Ruthenium(III) chloride, Ru content 45-55%
Cl3Ru