EMAIL THIS PAGE TO A FRIEND
Molecular & cellular proteomics : MCP
The first identification of lysine malonylation substrates and its regulatory enzyme.
PMID 21908771
Abstract
Protein post-translational modifications (PTMs) at the lysine residue, such as lysine methylation, acetylation, and ubiquitination, are diverse, abundant, and dynamic. They play a key role in the regulation of diverse cellular physiology. Here we report discovery of a new type of lysine PTM, lysine malonylation (Kmal). Kmal was initially detected by mass spectrometry and protein sequence-database searching. The modification was comprehensively validated by Western blot, tandem MS, and high-performance liquid chromatography of synthetic peptides, isotopic labeling, and identification of multiple Kmal substrate proteins. Kmal is a dynamic and evolutionarily conserved PTM observed in mammalian cells and bacterial cells. In addition, we demonstrate that Sirt5, a member of the class III lysine deacetylases, can catalyze lysine demalonylation and lysine desuccinylation reactions both in vitro and in vivo. This result suggests the possibility of nondeacetylation activity of other class III lysine deacetylases, especially those without obvious acetylation protein substrates. Our results therefore reveal a new type of PTM pathway and identify the first enzyme that can regulate lysine malonylation and lysine succinylation status.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P7634 3-Phosphoglyceric Phosphokinase from baker's yeast (S. cerevisiae), ammonium sulfate suspension, ≥1000 units/mg protein
A8811 Aldolase from rabbit muscle, ammonium sulfate suspension, 10-20 units/mg protein
A2714 Aldolase from rabbit muscle, lyophilized powder, ≥8.0 units/mg protein
E6126 Enolase from baker's yeast (S. cerevisiae), lyophilized powder, ≥50 units/mg protein
G5262 GAPDH, standard for protein electrophoresis
G5537 Glyceraldehyde-3-phosphate Dehydrogenase from baker's yeast (S. cerevisiae), lyophilized powder, 70-140 units/mg protein
G6019 Glyceraldehyde-3-phosphate Dehydrogenase from human erythrocytes, lyophilized powder, 50-150 units/mg protein
G2267 Glyceraldehyde-3-phosphate Dehydrogenase from rabbit muscle, lyophilized powder, ≥75 units/mg protein
M9004 Malic Dehydrogenase from bovine heart, ammonium sulfate suspension, 2000-4000 units/mg protein (modified Warburg-Christian)
M2634 Malic Dehydrogenase from porcine heart, buffered aqueous glycerol solution, 600-1000 units/mg protein (biuret)
M7383 Malic Dehydrogenase from porcine heart, ≥400 units/mg protein (biuret), ammonium sulfate suspension
M1567 Malic Dehydrogenase from porcine heart, ≥600 units/mg protein (biuret), ammonium sulfate suspension
N4773 Neuron-specific enolase from human brain, ≥95% (SDS-PAGE), buffered aqueous solution
P5538 Phosphoglucose Isomerase from Bacillus stearothermophilus, lyophilized powder, 300-1,000 units/mg protein
P5381 Phosphoglucose Isomerase from baker's yeast (S. cerevisiae), Type III, ammonium sulfate suspension, ≥400 units/mg protein (biuret)
F2668 Phosphoglucose Isomerase from baker's yeast (S. cerevisiae), for use with Fructose Assay Kit FA-20
P9544 Phosphoglucose Isomerase from rabbit muscle, Type XI, lyophilized powder, ≥200 units/mg protein