EMAIL THIS PAGE TO A FRIEND
Nature
Diffraction-unlimited all-optical imaging and writing with a photochromic GFP.
PMID 21909116
Abstract
Lens-based optical microscopy failed to discern fluorescent features closer than 200 nm for decades, but the recent breaking of the diffraction resolution barrier by sequentially switching the fluorescence capability of adjacent features on and off is making nanoscale imaging routine. Reported fluorescence nanoscopy variants switch these features either with intense beams at defined positions or randomly, molecule by molecule. Here we demonstrate an optical nanoscopy that records raw data images from living cells and tissues with low levels of light. This advance has been facilitated by the generation of reversibly switchable enhanced green fluorescent protein (rsEGFP), a fluorescent protein that can be reversibly photoswitched more than a thousand times. Distributions of functional rsEGFP-fusion proteins in living bacteria and mammalian cells are imaged at <40-nanometre resolution. Dendritic spines in living brain slices are super-resolved with about a million times lower light intensities than before. The reversible switching also enables all-optical writing of features with subdiffraction size and spacings, which can be used for data storage.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
92546 Abberior® CAGE 500, maleimide, for single-molecule switching microscopy (e.g. PALM, STORM, GSDIM)
C32H22F6N6O5
44254 Abberior® CAGE 500, NHS ester, for single-molecule switching microscopy (e.g. PALM, STORM, GSDIM)
C30H19F6N5O6
95705 Abberior® CAGE 532, maleimide, for single-molecule switching microscopy (e.g. PALM, STORM, GSDIM)
C38H34F6N6O7
92545 Abberior® CAGE 552, maleimide, for single-molecule switching microscopy (e.g. PALM, STORM, GSDIM)
C32H28N6O5
94822 Abberior® CAGE 552, NHS ester, for single-molecule switching microscopy (e.g. PALM, STORM, GSDIM)
C30H25N5O6
92544 Abberior® FLIP 565, maleimide, for single-molecule switching microscopy (e.g. PALM, STORM, GSDIM)
79189 Abberior® FLIP 565, NHS ester, for single-molecule switching microscopy (e.g. PALM, STORM, GSDIM)
38361 Abberior® STAR 440SXP, maleimide, for long Stokes STED and 2-color STED application
68221 Abberior® STAR 440SXP, NHS ester, for long Stokes STED and 2-color STED application
94716 Abberior® STAR 470SXP, NHS ester, for STED application
61048 Abberior® STAR 488, NHS ester, for STED application
03004 Abberior® STAR 512, maleimide, for STED application
38922 Abberior® STAR 512, NHS ester, for STED application
38377 Abberior® STAR 580, NHS ester, for STED application
96013 Abberior® STAR 635, maleimide, for STED application
30558 Abberior® STAR 635, NHS ester, for STED application
53366 Anti-Mouse IgG−Abberior® STAR 488 antibody produced in goat, for STED application
52944 Anti-Rabbit IgG−Abberior® STAR 488 antibody produced in goat, for STED application