EMAIL THIS PAGE TO A FRIEND
Zhongguo Zhong xi yi jie he za zhi Zhongguo Zhongxiyi jiehe zazhi = Chinese journal of integrated traditional and Western medicine
[Effect of cinobufacini injection on the tumor growth of tumor bearing rats of different constitutions].
PMID 21910344
Abstract
To observe effects of cold- and hot-property herbs and effects of hot and cold constitutions on the tumor growth of tumor bearing rats, and to observe the effect of Cinobufacini Injection (CI) on the tumor growth of tumor bearing rats of different constitutions. Eighty healthy male Wistar rats were randomly divided into eight groups, i.e., the tumor bearing control group, the tumor bearing heat syndrome group, the tumor bearing cold syndrome group, the heat syndrome tumor bearing group, the cold syndrome tumor bearing group, the tumor bearing CI group, the heat syndrome tumor bearing CI group, and the cold syndrome tumor bearing CI group, respectively. The weight and volume of rats' subcutaneous tumor were measured 14 days after tumor inoculation. The weight and volume of tumor in the heat syndrome tumor bearing CI group [(3.55 +/- 1.12) g, (2864.44 +/- 1430.51) mm3] and the tumor bearing CI group [(4.29 +/- 1.14) g, (3397.19 +/- 1701.13) mm3] were significantly lower than those of the tumor bearing control group [(6.01 +/- 2.45) g, (6218.91 +/- 3837.64) mm3] and the cold syndrome tumor bearing CI group [(6.90 +/- 1.57) g, (6168.42 +/- 2457.03) mm3], showing statistical difference (P<0.05). There was insignificant difference among other groups. CI showed better tumor inhibition effects on tumor bearing rats of heat syndrome constitution, which indicated CI was of cold property. It might be possibly used in tumor bearing rats of heat syndrome constitution.