EMAIL THIS PAGE TO A FRIEND
International endodontic journal
Comparison of ultrasonic irrigation and RinsEndo for the removal of calcium hydroxide and Ledermix paste from root canals.
PMID 21910743
Abstract
To compare the efficacy of ultrasonic irrigation and RinsEndo in removing calcium hydroxide and Ledermix paste from simulated root canal irregularities. The root canals of sixty extracted single-rooted teeth were prepared using FlexMaster rotary instruments to size 60, 0.02 taper. The roots were split longitudinally, and a standardized groove was prepared in the apical part of one segment. The teeth were randomly allocated into two groups (n = 30), according to the intracanal dressing. In the first group, grooves were filled with calcium hydroxide paste (Calxyl), whereas the grooves in the second group were filled with Ledermix paste. After reassembly, the root canals were completely filled with the respective medicament using a lentulo. The removal of medicament dressing was performed after 7 days with either passive ultrasonic irrigation or RinsEndo and 1% sodium hypochlorite for 3 min. The amount of remaining medicament was evaluated under a microscope with 30 × magnification using a four-grade scoring system. A regression analysis with P ≤ 0.05 was performed. Ledermix paste removal was significantly more effective than the removal of calcium hydroxide (P < 0.0001), whereas irrigation technique was not a significant factor (P = 0.3712). The percentages of complete removal of calcium hydroxide and Ledermix paste were 11.7% and 51.7%, respectively. None of the irrigation techniques was able to completely remove the intracanal medicaments from the apical part of the root canal. Irrespective of the irrigation technique, significantly less Ledermix paste was detected compared with calcium hydroxide.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D6140
Demeclocycline hydrochloride, powder or crystals
C21H21ClN2O8 · HCl
46161
Demeclocycline hydrochloride hydrate, VETRANAL®, analytical standard
C21H21ClN2O8 · HCl · xH2O