EMAIL THIS PAGE TO A FRIEND
Journal of clinical anesthesia
Prophylaxis of postoperative nausea and vomiting in elective breast surgery.
PMID 21911192
Abstract
To evaluate strategies to treat postoperative nausea and vomiting (PONV) in patients undergoing elective breast surgery. Prospective, randomized, double-blinded, placebo-controlled trial. University-affiliated hospital. 480 patients with risk factors for PONV. Patients were randomized to three groups to receive an antiemetic prophylactic combination of haloperidol and tropisetron (Group HT), dimenhydrinate and dexamethasone (Group DD), or no prophylaxis (Group P). Anesthesia was maintained with volatile anesthesia (desflurane or sevoflurane) and fentanyl or total intravenous anesthesia (TIVA). Incidence of nausea, emesis, or both in the early (0 - 2 hrs) and late (2 - 24 hrs) postoperative periods were recorded, as were the number of episodes and the time of each occurrence; and patient assessment of the PONV experience on a scale comparable to a numeric rating scale (NRS). Both antiemetic combinations significantly reduced PONV incidence. In patients who received no prophylaxis, PONV incidence was 48.2% in patients given volatile anesthetics and 43.8% in those who received TIVA. PONV incidence was 17.5% in the Group HT patients who received volatile anesthetics, and 25% in the Group HT patients who received TIVA. PONV incidence was 11.4% in Group DD patients given volatile anesthetics, and 15% in Group DD patients receiving TIVA. TIVA reduced the incidence of PONV in the early postoperative period (0-2 hrs), but increased PONV incidence in the late period (2-24 hrs). Patients given TIVA with propofol and remifentanil intraoperatively required more opioids postoperatively than patients given volatile anesthetics. The frequency of PONV was reduced significantly with both antiemetic combinations.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D2396
Dimenhydrinate
C17H21NO · C7H7ClN4O2