EMAIL THIS PAGE TO A FRIEND
Molecular bioSystems
Insights into the homocoupling reaction of 4-methylamino benzoic acid mediated by Trametes versicolor laccase.
PMID 21912806
Abstract
Spectroscopic measurements combined with Density Functional Theory calculations were applied to the characterization of the homocoupling reaction of 4-methylamino benzoic acid mediated by laccase.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
119695
4-(Methylamino)benzoic acid, 97%
C8H9NO2