EMAIL THIS PAGE TO A FRIEND
Journal of bacteriology
Whole genome sequence of the rifamycin B-producing strain Amycolatopsis mediterranei S699.
PMID 21914879
Abstract
Amycolatopsis mediterranei S699 is an actinomycete that produces an important antibiotic, rifamycin B. Semisynthetic derivatives of rifamycin B are used for the treatment of tuberculosis, leprosy, and AIDS-related mycobacterial infections. Here, we report the complete genome sequence (10.2 Mb) of A. mediterranei S699, with 9,575 predicted coding sequences.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
R0900000 Rifamycin B, European Pharmacopoeia (EP) Reference Standard
C39H49NO14