EMAIL THIS PAGE TO A FRIEND
Aviakosmicheskaia i ekologicheskaia meditsina = Aerospace and environmental medicine
[Structural and functional features of the rats' prefrontal cortex following 14-day suspension].
PMID 21916250
Abstract
Effects of 14-d suspension on the processes of rats' nervous cells conjugation in the brain cognitive cortex were studied. Signs of cognitive functions degradation and decrease of the number of conjugated nervous cells were found in the experimental group after suspension. This evidence are qualified as a negative effect of suspension on memory and physiological regeneration of neurons.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C5042
Cresyl Violet acetate, certified by the Biological Stain Commission
C18H15N3O3
255246
Cresyl Violet perchlorate
C16H11N3O · HClO4