EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry
5-Fluoro-4-thiouridine phosphoramidite: new synthon for introducing photoaffinity label into oligodeoxynucleotides.
PMID 21917468
Abstract
The synthesis of phosphoramidite of 5-fluoro-4-thio-2'-O-methyluridine is described. An appropriate set of protecting groups was optimized including the 4-thio function introduced via 4-triazolyl as the 4-(2-cyanoethyl)thio derivative, and the t-butyldimethyl silyl for 2' and 3' hydroxyl protection, enabling efficient synthesis of the phosphoramidite. These protecting groups prevented unwanted side reactions during oligonucleotide synthesis. The utility of the proposed synthetic route was proven by the preparation of several oligonucleotides via automated synthesis. Photochemical experiments confirmed the utility of the synthon.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T4509
4-Thiouridine, ≥98%
C9H12N2O5S