EMAIL THIS PAGE TO A FRIEND
Gene Therapy
A designed curved DNA sequence remarkably enhances transgene expression from plasmid DNA in mouse liver.
PMID 21918548
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
07-690
Anti-Histone H3 Antibody, CT, pan, serum, clone, Upstate®