EMAIL THIS PAGE TO A FRIEND
Angewandte Chemie (International ed. in English)
Molecular self-assembly of a homopolymer: an alternative to fabricate drug-delivery platforms for cancer therapy.
PMID 21919151
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N13038 Phosfolan, analytical standard