EMAIL THIS PAGE TO A FRIEND
Journal of the American Dental Association (1939)
New bitter blocker discovered, study finds.
PMID 21919220
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C11804
4-Sulfamoylbenzoic acid, 97%
C7H7NO4S
S0625
D-(−)-Salicin, ≥99% (GC)
C13H18O7