EMAIL THIS PAGE TO A FRIEND
Clinical journal of the American Society of Nephrology : CJASN
Attending rounds: patient with hypokalemia and metabolic acidosis.
PMID 21921151
Abstract
Hypokalemic paralysis represents a medical emergency requiring both rapid diagnosis and treatment. In this Attending Rounds a patient with hypokalemia and metabolic acidosis is presented to emphasize the role of routine laboratory studies in the assessment of such patients so that a correct diagnosis can be made and appropriate treatment can be initiated promptly.