EMAIL THIS PAGE TO A FRIEND
Rinsho shinkeigaku = Clinical neurology
[Familial amyloid polyneuropathy: diflunisal].
PMID 21921462
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D3281
Diflunisal, analytical standard
C13H8F2O3