EMAIL THIS PAGE TO A FRIEND
Shock
Pseudomonas aeruginosa is associated with increased lung cytokines and asymmetric dimethylarginine compared with methicillin-resistant Staphylococcus aureus.
PMID 21921834