EMAIL THIS PAGE TO A FRIEND
Chemistry (Weinheim an der Bergstrasse, Germany)
A molecular probe for the highly selective chromogenic detection of DFP, a mimic of Sarin and Soman nerve agents.
PMID 21922586
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D0879
Diisopropylfluorophosphate
C6H14FO3P