EMAIL THIS PAGE TO A FRIEND
ChemSusChem
Ruthenium-catalyzed conversion of levulinic acid to pyrrolidines by reductive amination.
PMID 21922683
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
W262706
Levulinic acid, ≥97%, FG
C5H8O3
W262701
Levulinic acid, natural, 99%, FG
C5H8O3