EMAIL THIS PAGE TO A FRIEND
Journal of the European Academy of Dermatology and Venereology : JEADV
Lacking CD56 expression in a relapsing cutaneous blastic plasmacytoid dendritic cell neoplasm after allogeneic bone marrow transplantation: FISH analysis revealed loss of 11q.
PMID 21923812