EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry
Potent inhibition of Norwalk virus by cyclic sulfamide derivatives.
PMID 21925886
Abstract
A new class of compounds that exhibit anti-norovirus activity in a cell-based system and embody in their structure a cyclosulfamide scaffold has been identified. The structure of the initial hit (compound 2a, ED(50) 4 μM, TD(50) 50 μM) has been prospected by exploiting multiple points of diversity and generating appropriate structure-activity relationships.