EMAIL THIS PAGE TO A FRIEND
Doklady. Biochemistry and biophysics
Modified nanodiamonds obtained by detonation synthesis in constructing biochemical indication systems (As exemplified by the glucose determination system).
PMID 21928140