EMAIL THIS PAGE TO A FRIEND
Journal of nephrology
Podocyte Wnt/ß-catenin pathway is activated by integrin-linked kinase in clinical and experimental focal segmental glomerulosclerosis.
PMID 21928230
Abstract
Changes in podocyte phenotype and function are characteristic of proteinuric glomerular diseases. Integrin-linked kinase (ILK) functions as a common downstream effector in proteinuric diseases. In addition, ILK was shown to interact with the Wnt signaling pathway. Here, we investigated ILK expression as well as its involvement with the Wnt signaling pathway in renal biopsies of patients with primary focal segmental glomerulosclerosis (FSGS), and in a correspondent in vivo model of podocyte lesion. Biopsies from 37 patients with primary FSGS were evaluated by immunohistochemistry for ILK, phosphorylated GSK-3ß (pGSK-3ß) and ß-catenin expression. As experimental model, male Wistar rats received 5 injections of puromycin aminonucleoside (PAN) at 2-week intervals, and their kidneys were evaluated for ILK, P-cadherin and pAkt expression as well as ß-catenin and LEF-1 colocalization. Patients presented de novo ILK expression and pGSK-3ß in podocytes. In animals, there was an increase in gene and protein expression of ILK, mainly detected in the podocytes, as well as increased protein expression of pAkt compared with controls. ß-Catenin translocated to the nuclei of podocytes in animals and patients. ß-Catenin colocalized with LEF-1 in the nuclei of podocytes of animals. Gene expression of ß-catenin and P-cadherin in PAN rats was lower compared with controls. Our findings suggest that activation of ILK activated the Wnt signaling pathway in damaged podocytes. This phenomenon could have an important role in development and/or progression of clinical and experimental FSGS.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P7130
Puromycin aminonucleoside
C12H18N6O3