EMAIL THIS PAGE TO A FRIEND
Angewandte Chemie (International ed. in English)
Shape-controlled synthesis of copper nanocrystals in an aqueous solution with glucose as a reducing agent and hexadecylamine as a capping agent.
PMID 21928444
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
445312
Hexadecylamine, 98%
C16H35N
H7408
Hexadecylamine, technical grade, 90%
C16H35N