EMAIL THIS PAGE TO A FRIEND
Archives of dermatology
Dermatitis to FD&C yellow No. 6 dye in orange antiseptic solution.
PMID 21931066
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
465224
Sunset Yellow FCF, Dye content 90 %
C16H10N2Na2O7S2
68775
Sunset Yellow FCF, analytical standard
C16H10N2Na2O7S2