EMAIL THIS PAGE TO A FRIEND
Circulation
Four-limb acute ischemia induced by ergotamine in an AIDS patient treated with protease inhibitors.
PMID 21931103
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
45510
Ergotamine D-tartrate, ≥97.0% (calc. based on dry substance, NT)
C33H35N5O5 · 0.5C4H6O6
45512
Ergotamine D-tartrate, tested according to Ph.Eur.
C33H35N5O5 · 0.5C4H6O6