EMAIL THIS PAGE TO A FRIEND
Organic letters
Phosphine-initiated general base catalysis: facile access to benzannulated 1,3-diheteroatom five-membered rings via double-Michael reactions of allenes.
PMID 21932819
Abstract
General base-catalyzed double-Michael reactions of allenes with various dinucleophiles are described. The reactions are facilitated most efficiently by a catalytic amount of trimethylphosphine, affording six types of C2-functionalized benzannulated five-membered heterocycles: benzimidazolines, benzoxazolines, benzothiazolines, 1,3-benzodioxoles, 1,3-benzoxathioles, and 1,3-benzodithioles. This atom-economical reaction is operationally simple and provides the product heterocycles in good to excellent yields. Careful mechanistic studies unveiled the phosphine-triggered general base catalysis pathway. Furthermore, the double-Michael reaction can serve as an alternative method for the selective monoketalization of β-diketones.