EMAIL THIS PAGE TO A FRIEND
Peptides
Gastrin-releasing peptide induces itch-related responses through mast cell degranulation in mice.
PMID 21933692
Abstract
Gastrin-releasing peptide (GRP), secreted from the central terminals of primary afferents, is involved in the transmission of itch signals in the spinal dorsal horn. Although primary afferents containing GRP are distributed throughout the skin, the role of peripherally released GRP in the itch response is unknown. We investigated whether GRP acts on the skin to induce an itch response in mice. Intradermal injections of GRP(18-27) (1-300 nmol/site) elicited scratching. GRP(18-27)-induced scratching was inhibited by the μ-opioid receptor antagonist naltrexone hydrochloride, the BB(2) bombesin receptor antagonist RC-3095, the H(1) histamine receptor antagonists fexofenadine hydrochloride and chlorpheniramine maleate, and the PAR(2) proteinase-activated receptor antagonist FSLLRY-NH(2). Mast cell deficiency significantly, but not completely, reduced the GRP(18-27)-induced scratching. BB(2) bombesin receptors are present in mast cells in the skin, and intradermal injection of GRP(18-27), not only induced scratching, but also led to mast cell degranulation. GRP(18-27)-induced mast cell degranulation was inhibited by the BB(2) bombesin receptor antagonist RC-3095. These results suggest that peripherally released GRP can induce an itch response, at least partly, through activation of BB(2) receptors present in the mast cells, triggering their degradation and the release of histamine and the serine proteinase, tryptase.