EMAIL THIS PAGE TO A FRIEND
Bioresource technology
Degradation of the antibiotics norfloxacin and ciprofloxacin by a white-rot fungus and identification of degradation products.
PMID 21937221
Abstract
More than 90% of the antibiotics ciprofloxacin (CIPRO) and norfloxacin (NOR) at 2 mg L(-1) were degraded by Trametes versicolor after 7 days of incubation in malt extract liquid medium. In in vitro assays with purified laccase (16.7 nkat mL(-1)), an extracellular enzyme excreted constitutively by this fungus, 16% of CIPRO was removed after 20 h. The addition of the laccase mediator 2,2-azino-bis-(3-ethylbenzthiazoline-6-sulfonic acid) diammonium salt led to 97.7% and 33.7% degradation of CIPRO and NOR, respectively. Inhibition of CIPRO and NOR degradation by the cytochrome P450 inhibitor 1-aminobenzotriazole suggests that the P450 system also plays a role in the degradation of the two antibiotics. Transformation products of CIPRO and NOR were monitored at different incubation times by triple-quadrupole and quadrupole time-of-flight mass spectrometry, and can be assigned to three different reaction pathways: (i) oxidation of the piperazinyl substituent, (ii) monohydroxylation, and (iii) formation of dimeric products.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A3940
1-Aminobenzotriazole
C6H6N4