EMAIL THIS PAGE TO A FRIEND
Organic & biomolecular chemistry
Stereoselective preparation of β,γ-methano-GABA derivatives.
PMID 21938296
Abstract
The Kulinkovich-de Meijere reaction between an unsaturated Grignard reagent and a chiral amide takes place with a high trans stereoselectivity and provides a convenient access to non-racemic trans cyclopropylamines. These compounds are transformed in four steps into the corresponding N-protected β,γ-methano-GABA derivatives, which are obtained for the first time in enantiomerically pure form. The corresponding transformations of the cis cyclopropylamine adducts are also described.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
125504
Cyclopropylamine, 98%
C3H7N