EMAIL THIS PAGE TO A FRIEND
Forensic science international
Determination of deslanoside in antemortem and postmortem specimens. Unusual case report.
PMID 2193860
Abstract
One case of the erroneous administration of deslanoside and high level of drug in antemortem plasma and postmortem specimens has been reported owing to the unusual surrounding circumstances. Deslanoside in antemortem plasma was determined by FPIA and the analysis was done by HPLC in the postmortem tissue samples. The analytical results and methods used in the examinations are discussed in the following paper.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D0600000 Deslanoside, European Pharmacopoeia (EP) Reference Standard
C47H74O19