EMAIL THIS PAGE TO A FRIEND
Journal of medicinal chemistry
Water-soluble renin inhibitors: design of a subnanomolar inhibitor with a prolonged duration of action.
PMID 2194033
Abstract
Incorporation of nonreactive polar functionalities at the C- and N-termini of renin inhibitors led to the development of a subnanomolar compound (21) with millimolar solubility. This inhibitor demonstrated excellent efficacy and a long duration of action upon intravenous administration to monkeys. While activity was also observed intraduodenally, a comparison of the blood pressure responses indicated low bioavailability. Subsequent experiments in rats showed that, although the compound was absorbed from the gastrointestinal tract, extensive liver extraction severely limited bioavailability.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
377112
3-Pyrroline, 95%
C4H7N